Learn More
Medchemexpress LLC T3-ATA S-isomer 100mg | 2438721-48-1 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY114271A100MG
Encompass_Preferred
T3-ATA (S-isomer) is the S-stereoisomer of T3-ATA, a thyroid hormone analog used as a research reagent to probe thyroid hormone receptor function. It is supplied for laboratory research use only and is not intended for human or clinical use.
  • Research reagent for thyroid hormone receptor assays.
  • S-stereoisomer (active stereochemistry) for receptor selectivity studies.
  • Molecular weight 767.11; formula C19H16I3NO6S.
  • Store protected from light and under inert atmosphere; follow supplier storage recommendations.
  • For in vitro pharmacology, binding, and biochemical studies.
Catalog No. 50-002-28576