Learn More
Apexbio Technology LLC HyperScribe™ T3 High Yield RNA Synthesis Kit 100rxns
SDP
Supplier:  Apexbio Technology LLC K1416100
Encompass_Preferred
HyperScribe T3 High Yield RNA Synthesis Kit is designed for in vitro transcription based on the activity of T3 RNA polymerase The kit contains T3 RNA polymerase optimized reaction buffer ribonucleoside triphosphates (ATP CTP GTP UTP) with high purity and defined concentration and control DNA template Utilizing these NTPs this system synthesizes RNA transcripts suitable for radiolabeled RNA probes and RNA modifications including capped dye-labeled or biotinylated nucleotides A standard 20 L reaction typically yields 50 g unmodified RNA from 1 g template DNA The synthesized RNA is applicable for in vitro translation antisense RNA studies RNA interference (RNAi) RNA vaccines RNA structure and function analysis ribozyme biochemistry RNase protection assays and hybridization-based blotting techniques
Catalog No. 50-000-64303