Learn More
Bulbman Inc FCS 64640HLX 24V Osram 54263
SDP
Supplier:  Bulbman Inc 104409
Encompass_Preferred
Sylvania 54263 HXL lamp has a T5 bulb shape and a G6.35 base type. It operates with 150 watts and 24 volts and will typically last an average of 50 hours. AKA: 64640HLX and A1/216
Catalog No. NC9652191