Learn More
Medchemexpress LLC T56-LIMKi | 924473-59-6 | 98.7% | 389.33 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY19352100MG
Encompass_Preferred
T56-LIMKi is a selective inhibitor of LIMK2 that efficiently inhibits the growth of various cancer cell lines including Panc-1, ST88-14, U87, and A549 lung cancer cells. It operates by decreasing phosphorylated cofilin levels, thereby inhibiting tumor cell migration, growth, and anchorage-independent colony formation.
  • Selective inhibitor of LIMK2
  • Inhibits the growth of various cancer cell lines
  • Decreases phosphorylated cofilin levels
  • Blocks cofilin phosphorylation, leading to actin severance
  • Reduces tumor cell migration and growth
  • Induces tumor shrinkage in vivo in mouse models
Catalog No. 50-003-85057