Learn More
Apexbio Technology LLC HyperScribe™ T7 High Yield RNA Synthesis Kit, 50 rxns
SDP
Supplier:  Apexbio Technology LLC K104750
Encompass_Preferred
HyperScribe™ T7 High Yield RNA Synthesis Kit performs in vitro RNA transcription experiment in a relatively short time using T7 RNA polymerase. The kit is designed for high yields of RNA transcripts, highly specific radiolabeled RNA probes and to obtain capped, dye-labeled or biotinylated RNA incorporated modified nucleotides. RNA synthesized using this kit covers a wide range of applications, such as in vitro translation, antisense RNA and RNAi experiments, RNA vaccines, RNA structure and function studies, ribozyme biochemistry, RNase protein experiments and probe-based hybridization blots. The kit contains sufficient reagents to carry out 25/50/100 reactions, 20 l each time. Up to 180 g of RNA can be generated with 1 g of control template per standard reaction. Each 25/50/100 reactions kit can produce up to 4.5 mg/9 mg/18 mg RNA. Other sizes are also available. Please inqury us for quote.
Catalog No. 50-199-8279