Learn More
CPC Scientific H-Thr-Met-Pro-Phe-Leu-Phe-Cys-Asn-Val-Asn-Asp-Val-Cys-Asn-Phe-Ala-Ser-Arg-Asn-Asp-Tyr-Ser-Tyr-Trp-Leu-OH (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific CASP093B
Encompass
Tumstatin is an endothelial cell-specific inhibitor of protein synthesisONE-LETTER SEQUENCE: TMPFLFCNVNDVCNFASRNDYSYWL (C7&C13 bridge)MOLECULAR FORMULA: C137H189N33O39S3MOLECULAR WEIGHT:3018.4STORAGE CONDITIONS: -20 5CRESEARCH AREA: ApoptosisREFERENCES: Maeshima Y., et al., Science 295, 140-143 (2002)
Catalog No. 50-210-9097