Learn More
Selleck Chemical LLC T863 S0359-25MG
SDP
Supplier:  Selleck Chemical LLC S035925MG
Encompass_Preferred
T863 (DGAT-3) is a potent selective and orally active inhibitor of Acyl-CoA diacylglycerol acyltransferase 1 (DGAT1) tht acts on the acyl-CoA binding site of DGAT1 and inhibits DGAT1-mediated triacylglycerol formation in cells
Catalog No. 50-000-42310