Learn More
TARGETMOL CHEMICALS INC TABERNANTHALOG 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T602215MG
Encompass_Preferred
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Tabernanthalog (TBG) is a 5-HT2A agonist. In rodentsTabernanthalog has been found to promote structural neuroplasticity reduce alcohol-seeking and heroin-seeking behavior and produce antidepressant effects. purity: 98%
Catalog No. 50-257-9632