Learn More
Selleck Chemical LLC Tabersonine hydrochloride S5101-25mg
SDP
Supplier:  Selleck Chemical LLC S510125MG
Encompass_Preferred
Tabersonine an ingredient extracted from the bean of Voacanga africana is a potent inhibitor against A (1 42) aggregation and toxicity
Catalog No. 50-311-2329