Learn More
Medchemexpress LLC Tacalcitol monohydrate | 93129-94-3 | 99.9% | 434.65 | 1 MG

Supplier: Medchemexpress LLC HY323381MG
Tacalcitol monohydrate can promote normal bone growth by regulating calcium ions. It inhibits cancer cell proliferation and migration and can be used in the research of inflammation, cancer, and skin diseases. This product is for research use only and not sold to patients.
- Purity: 99.85%
- Appearance: Solid
- Color: White to off-white
- Shipping: Room temperature in continental US; may vary elsewhere
- Storage: 4°C, protect from light, stored under nitrogen. In solvent: -80°C, 6 months; -20°C, 1 month (protect from light, stored under nitrogen).
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.