Learn More
Medchemexpress LLC Tacalcitol monohydrate | 93129-94-3 | MFCD13184757 | 99.9% | 434.65 g/mol | C27H46O4 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY3233810MG
Encompass_Preferred
Tacalcitol monohydrate is a vitamin D3 analog (1,24(R)-dihydroxyvitamin D3 monohydrate) used as a research reagent to modulate vitamin D receptor (VDR) signaling and calcium regulation. It is applied in studies of bone metabolism, skin disorders, inflammation, and oncology to probe VDR-mediated pathways.
  • High purity (99.85%).
  • Vitamin D3 analog that modulates vitamin D receptor activity.
  • Applicable to in vitro and in vivo research on bone metabolism, skin disease, inflammation, and cancer.
  • Supplied as a monohydrate solid for consistent dosing.
  • Molecular weight 434.65 g/mol; formula C27H46O4.
Catalog No. 50-002-97597