Learn More
Cayman Chemical TaccalonlIde A 10mg
SDP
Supplier:  Cayman Chemical 4079810 MG
Encompass_Preferred
A taccalonolide with anticancer activity; induces microtubule bundling, formation of abnormal mitotic spindles, and cell cycle arrest at the G2/M phase in HeLa cells at 3.5 µM; decreases the proliferation of HeLa cells (IC50 = 594 nM); increases the number of tumor free mice but reduces body weight in a 16/C murine breast cancer model
Catalog No. 50-294-3566