Learn More
Selleck Chemical LLC Tacrine E4822-100MG
SDP
Supplier:  Selleck Chemical LLC E4822100MG
Encompass_Preferred
Tacrine is a potent acetylcholinesterase (AChE) inhibitor It inhibits acetylcholinesterase(AChE) and butyrylcholinesterase(BChE) with an IC50 of 77 nM and 69 nM respectively It is used in the treatment of Alzheimer s disease (AD) also poisoning topoisomerases and inhibiting in vivo (mitochondrial)mtDNA synthesis
Catalog No. 50-000-43535