Learn More
Apexbio Technology LLC Tacrine hydrochloride 1684-40-8 10mM (in 1mL H2O)
SDP
Supplier:  Apexbio Technology LLC B65275.1
Encompass_Preferred
Tacrine hydrochloride (CAS 1684-40-8) is a small molecule inhibitor of cholinesterases exhibiting potent inhibitory activity against both acetylcholinesterase (AChE IC 31 nM) and butyrylcholinesterase (BChE IC 25 6 nM) in vitro Additionally it has been shown to inhibit N-methyl-D-aspartate receptors (NMDARs) with an IC of approximately 26 M By modulating both cholinergic and glutamatergic signaling pathways tacrine hydrochloride is frequently employed in studies investigating the molecular mechanisms underlying Alzheimer s disease and exploring potential therapeutic strategies for neurodegenerative disorders
Catalog No. 50-000-92034