Learn More
Selleck Chemical LLC Tacrine hydrochloride hydrate S3639-1g
SDP
Supplier:  Selleck Chemical LLC S36391G
Encompass_Preferred
Tacrine (Tetrahydroaminacrine Tetrahydroaminoacridine) is a centerally active cholinesterase inhibitor that has been used to counter the effects of muscle relaxants as a respiratory stimulant and in the treatment of Alzheimer s disease and other central nervous system disorders It also acts as a histamine N-methyltransferase inhibitor
Catalog No. 50-310-9753