Learn More
TARGETMOL CHEMICALS INC Tacrine hydrochloride hydrate
SDP
Supplier:  TARGETMOL CHEMICALS INC T2191500MG
Encompass_Preferred
Also available in 1 mL, 100 mg, 200 mg and bulk. Please contact Fisher for quotes. Tacrine hydrochloride hydrate (Tetrahydroaminacrine) is a cholinesterase inhibitor that crosses the blood-brain barrier. Tacrine has been used to counter the effects of muscle relaxants, as a respiratory stimulant, and in the treatment of Alzheimer's disease and other central nervous system disorders.
Catalog No. 50-287-9555