Learn More
Medchemexpress LLC HY-13756 200mg , Tacrolimus CAS:104987-11-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY13756200MG
Encompass_Preferred
HY-13756 200mg Tacrolimus CAS: 104987-11-3 Tacrolimus (FK506), a macrocyclic lactone, binds to FK506 binding protein (FKBP) to form a complex. Tacrolimus inhibits calcineurin phosphatase, which inhibits T-lymphocyte signal transduction and IL-2 transcription. Immunosuppressive properties. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0101