Learn More
Selleck Chemical LLC Tacrolimus (FK506) 10mM/1mL 104987-11-3 FR900506, Fujimycin, Prograf
SDP
Supplier:  Selleck Chemical LLC S500310MM/1ML
Encompass_Preferred
Tacrolimus (FK506, FR900506, Fujimycin, Prograf) is a 23-membered macrolide lactone, it reduces peptidyl-prolyl isomerase activity in T cells by binding to the immunophilin FKBP12 (FK506 binding protein) creating a new complex. Tacrolimus also inhibits the phosphatase activity of calcineurin. Tacrolimus induces vascular endothelial autophagy. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-4305