Learn More
Medchemexpress LLC Tacrolimus (Standard) | 104987-11-3 | 99.6% | 804.02 | 50 MG
Supplier: Medchemexpress LLC HY13756R50MG
Tacrolimus (Standard) is the analytical standard of Tacrolimus, intended for research and analytical applications. It functions as a macrocyclic lactone that binds to FK506 binding protein (FKBP) to form a complex. Tacrolimus inhibits calcineurin phosphatase, which then inhibits T-lymphocyte signal transduction and IL-2 transcription, ultimately possessing immunosuppressive properties. It is suitable for a variety of research assays and experiments.
- Analytical standard grade
- Reference standard for assays
- Used in qualitative, quantitative, and methodological research experiments
- Compatible with techniques such as HPLC, GC, and MS