Learn More
Apexbio Technology LLC Tadalafil, 100mg. Cas: 171596-29-5 MFCD: MFCD07771966. PDE5 inhibitor
SDP
Supplier:  Apexbio Technology LLC A4327100
Encompass_Preferred
Tadalafil, also known as IC351 or Cialis, is a potent, reversible and competitive small-molecule inhibitor of phosphodiesterase 5 (PDE5). 171596-29-5. MFCD07771966
Catalog No. 50-101-2624