Learn More
Strem, An Ascensus Company CAS# 952649-50-2. 25mg. (3aR,8aR)-Tetrahydro-6-hydroxy-2,2-dimethyl-4,4,8,8-tetra-2-naphthalenyl-6-oxide-1,3-dioxolo[4,5-e][1,3,2]dioxaphosphepin, 98%, (99% ee).
SDP
Supplier:  Strem, An Ascensus Company 15860225MG
Encompass
CAS# 952649-50-2. 25mg. (3aR,8aR)-Tetrahydro-6-hydroxy-2,2-dimethyl-4,4,8,8-tetra-2-naphthalenyl-6-oxide-1,3-dioxolo[4,5-e][1,3,2]dioxaphosphepin, 98%, (99% ee). . Molecular Weight: 728.8. Molecular Formula: C47H37O6P. Color/form: off white to light yellow pwdr. Strem# 15-8602. http://www.strem.com/catalog/v/15-8602/
Catalog No. 50-206-5502