Learn More
Alkali Scientific TAE 10X Solution [TRIS Acetate EDTA 10X Solution], 10 Liters
Supplier: Alkali Scientific TAE003CP
TAE 10X Solution is a ready-to-use buffer for electrophoresis applications, specifically designed for DNA analysis. The 10-liter solution is concentrated to 10X strength, making it highly efficient for routine gel electrophoresis, particularly for the separation of large DNA fragments. This buffer contains TRIS, acetate, and EDTA, which help maintain the stability of DNA during electrophoresis by preventing degradation. The 10X concentration allows users to easily dilute the buffer to a working strength, reducing preparation time and cost. TAE is widely used in laboratories for DNA analysis, PCR product analysis, and DNA sequencing applications, ensuring high resolution and reproducibility in gel electrophoresis.