Learn More
Selleck Chemical LLC TAE226 (NVP-TAE226) 5mg 761437-28-9
SDP
Supplier:  Selleck Chemical LLC S28205MG
Encompass_Preferred
TAE226 (NVP-TAE226) is a potent FAK inhibitor with IC50 of 5.5 nM and modestly potent to Pyk2, ~10- to 100-fold less potent against InsR, IGF-1R, ALK, and c-Met. TAE226 (NVP-TAE226) induces apoptosis. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-3019