Learn More
Cayman Chemical TAF 10 Peptide Other Proteins
SDP
Supplier:  Cayman Chemical 102281 EA
Encompass_Preferred
TAF10 is one of many protein factors or coactivators associated with RNA polymerase II activity. One vial of this peptide may be used as a methyltransferase acceptor peptide for more than 200 reactions 15 µM.
Catalog No. 50-288-6762