Learn More
TARGETMOL CHEMICALS INC TAFAMIDIS MEGLUMINE 25MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T758225MG
Encompass_Preferred
Also available in 5 mg 10 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Tafamidis meglumine (Fx-1006A) is a Potent and Selective Transthyretin Kinetic Stabilizer That Inhibits the Amyloid Cascadepurity: 99%
Catalog No. 50-257-9179