Learn More
Selleck Chemical LLC Tafamidis meglumine E4852-100MG
SDP
Supplier:  Selleck Chemical LLC E4852100MG
Encompass_Preferred
Tafamidis meglumine (Vyndaqel Fx-1006A) stabilizes the tetrameric structure of transthyretin (TTR) by binding to thyroxine-binding sites Tafamidis meglumine treats transthyretin familial amyloid polyneuropathies (TTR-FAP) a condition characterized by neurological damage from TTR amyloidosis by preventing the misfolding and aggregation of transthyretin (TTR) into amyloid fibrils
Catalog No. 50-000-41915