Learn More
Medchemexpress LLC Tafamidis meglumine | 951395-08-7 | 99.8% | 503.33 | 5 MG
SDP
Supplier:  Medchemexpress LLC 5MGHY14852A
Encompass_Preferred
Tafamidis meglumine is a potent and selective transthyretin (TTR) stabilizer. It shows comparable potency and efficacy to the mutant homotetramers V30M-TTR, V122I-TTR, and wild type WT-TTR, with EC50s of 2.7-3.2 μM. Tafamidis meglumine inhibits amyloidogenesis.
  • Potent and selective transthyretin (TTR) stabilizer
  • Inhibits amyloidogenesis
  • Comparable potency and efficacy to mutant homotetramers V30M-TTR, V122I-TTR, and wild type WT-TTR
  • Binds selectively to thyroxine-binding sites of the tetramer
  • Kinetically stabilizes TTR
Catalog No. 50-003-64330