Learn More
Cayman Chemical Tafluprost-d4 1mg
SDP
Supplier:  Cayman Chemical 90024051 MG
Encompass_Preferred
An internal standard for the quantification of tafluprost by GC- or LC-MS
Catalog No. 50-289-3542