Learn More
Cayman Chemical Tafluprostfree acId-d4 1mg
SDP
Supplier:  Cayman Chemical 90024061 MG
Encompass_Preferred
An internal standard for the quantification of tafluprost (free acid) by GC-or LC-MS
Catalog No. 50-289-2714