Learn More
Cayman Chemical Tafluprostfree acId-d4 50UG
SDP
Supplier:  Cayman Chemical 900240650 UG
Encompass_Preferred
An internal standard for the quantification of tafluprost (free acid) by GC-or LC-MS
Catalog No. 50-289-4286