Learn More
Diagenode Inc Tagmentase TN5 TRN Unloaded, 10 μl, Hyperactive Tn5 transposase, Mg++ dependent, 19-bp ME DNA sequence required, 2 mg/ml protein concentration, 53.3 kDa molecular weight
Supplier: Diagenode Inc C0107001010
Diagenode Tagmentase (Tn5 transposase) is a hyperactive enzyme designed for a wide range of molecular biology applications. It enables an efficient "cut and paste" tagmentation reaction, allowing simultaneous DNA cleavage and the insertion of specific oligonucleotides or sequencing adapters. As an unloaded variant, it provides researchers with the flexibility to customize the enzyme with oligonucleotides tailored to their specific experimental needs. This Tn5 transposase is essential for various Next-Generation Sequencing workflows.
- Enzyme type: Hyperactive Tn5 transposase
- Loading status: Unloaded
- Function: Catalyzes "cut and paste" tagmentation
- Activity requirement: Mg++ dependent
- Optimal pH: 7.5-8.0
- Optimal temperature: 37-55 °C
- Storage conditions: Store at -20 °C
- Protein concentration: 2 mg/ml
- Volume: 10 μl
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.