Learn More
Selleck Chemical LLC TAI-1
SDP
Supplier:  Selleck Chemical LLC S74955MG
Encompass_Preferred
TAI-1 is a potent and specific Hec1 inhibitor which disrupts Hec1-Nek2 protein interaction
Catalog No. 50-217-3278