Learn More
TARGETMOL CHEMICALS INC TAI-1 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T66915MG
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. TAI-1 is a potent and specific Hec1 inhibitor which disrupts Hec1-Nek2 protein interaction. purity: 98%
Catalog No. 50-257-9305