Learn More
Med Vet International Tail Docking Instrument, 8-1/2in, J0176, Each
Supplier:  Med Vet International JOR176
Encompass_Preferred
Great instrument for getting that special cut on docking puppies tails. The special curved blade cuts clean and leaves an extra portion of skin. A simple figure 8-suture pattern in absorbable suture gives a very cosmetic result. Stainless steel with brass cutting board, 81.2"L
Catalog No. 50-005-85050