Learn More
Braintree Scientific Tail Vein Injection Platform
SDP
Supplier:  Braintree Scientific IL300
Encompass
Tail Vein Injection Platform - The quickest and easiest restraint for tail vein injections. Simply pull mouse tail through the slot and make injection! Features Non-grip surface, Wide opening for easy access, Open slot for easy cleaning - feces and urine easily washed away. **Sold for research purposes only and not for human use**
Catalog No. NC9963133