Learn More
Cayman Chemical TAK-242, 243984-11-4, 10 mg
SDP
Supplier:  Cayman Chemical 1387110
Encompass_Preferred
Molecular Formula C15H17NO4ClFS, Purity ≥98%, Formula Weight 361.8, Synonyms: Resatorvid; A cell-permeable inhibitor of TLR4 signaling, blocking LPS-induced production of NO, TNF-, IL-6, and IL-1 in macrophages with IC50 values of 1-11 nM; effective in vivo, suppressing the production of NO, TNF-, IL-6, and IL-1 in mice treated with LPS when given intravenously.
Catalog No. NC1535851