Learn More
Apexbio Technology LLC TAK-242, 243984-11-4, MFCD11046362, 5 mg
SDP
Supplier:  Apexbio Technology LLC A38505
Encompass_Preferred
MF: C15H17ClFNO4S, MW: 361.82, Solubility: 18.091 mg/mL in DMSO, 100.6 mg/mL in EtOH,insoluble in H2O. With IC50 of 1.1 to 11 nM, TAK-242 inhibited LPS-induced NO production, tumor necrosis factor-alpha and interleukin (IL)-6 in RAW264.7 cells and mouse peritoneal macrophages.
Catalog No. 50-115-0780