Learn More
Medchemexpress LLC HY-117959 5mg Medchemexpress, TAK-615 CAS:1664335-55-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1179595MG
Encompass_Preferred
Medchemexpress, HY-117959 5mg TAK-615 CAS:1664335-55-0 TAK-615 is a negative allosteric modulator (NAM) of the LPA1 receptor for the research of pulmonary fibrosis. TAK-615 binds the LPA1 receptor with high affinity (Kd high affinity of 1.7 nM and Kd low affinity of 14.5 nM). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-0911