Learn More
Medchemexpress LLC HY-15767 50mg , TAK-632 CAS:1228591-30-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1576750MG
Encompass_Preferred
HY-15767 50mg TAK-632 CAS: 1228591-30-7 TAK-632 is a potent pan-RAF inhibitor with IC50 of 1.4, 2.4 and 8.3 nM for CRAF, BRAFV600E, BRAFWT, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0174