Learn More
TARGETMOL CHEMICALS INC TAK-653 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T615185MG
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. TAK-653 a selective positive allosteric modulator (PAM) of AMPA receptors exhibiting minimal agonistic activity elicits an antidepressant-like response while maintaining a favorable safety profile in rat models. purity: 99%
Catalog No. 50-257-9516