Learn More
Selleck Chemical LLC TAK-715 10mg 303162-79-0
SDP
Supplier:  Selleck Chemical LLC S292810MG
Encompass_Preferred
TAK-715 is a p38 MAPK inhibitor for p38? with IC50 of 7.1 nM, 28-fold more selective for p38? over p38?, no inhibition to p38?/?, JNK1, ERK1, IKK?, MEKK1 or TAK1. Phase 2. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-3232