Learn More
Medchemexpress LLC Tak-901 | 934541-31-8 | 99.8% | 50 MG
SDP
Supplier:  Medchemexpress LLC HY1220150MG
Encompass_Preferred
TAK-901 is a multi-targeted Aurora kinase inhibitor used in research to inhibit Aurora A and Aurora B, with reported IC50 values of 21 nM and 15 nM, respectively. It is supplied as a high-purity powder suitable for in vitro and preclinical studies.
  • Inhibits Aurora A (IC50 = 21 nM) and Aurora B (IC50 = 15 nM).
  • High purity for reliable assay and screening results.
  • Powder form allows flexible preparation and dosing.
  • Recommended storage preserves stability for extended use.
Catalog No. 50-002-30101