Learn More
TARGETMOL CHEMICALS INC TAK-960 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T72005MG
Encompass_Preferred
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. TAK-960 is an orally bioavailable selective inhibitor of Plks with IC50 values of 0.8 16.9 and 50.2 nM for Plk1 Plk2 and Plk3 respectively. purity: 98%
Catalog No. 50-257-6335