Learn More
Medchemexpress LLC Tak1-in-2 | 2763213-98-3 | 98.8% | C26H26F2N6O5 | 10MG
SDP
Supplier:  Medchemexpress LLC HY13217210MG
Encompass_Preferred
TAK1-IN-2 is a potent, selective small-molecule inhibitor of transforming growth factor-β-activated kinase 1 (TAK1) for laboratory research. It exhibits low-nanomolar potency (IC50 = 2 nM) and is supplied as a high-purity compound for in vitro and preclinical studies; not for human or clinical use.
  • Potent TAK1 inhibition (IC50 = 2 nM).
  • High selectivity reduces off-target kinase activity.
  • High purity suitable for biochemical and cell-based assays.
  • Available in multiple milligram pack sizes for dosing flexibility.
  • Datasheet, certificate of analysis, and safety data sheet provided.
  • Intended for research use only; not for human therapeutic use.
Catalog No. 50-002-59892