Learn More
TARGETMOL CHEMICALS INC TALABOSTAT MESYLATE 10MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T404210MG
Encompass_Preferred
Also available in 1 mg 5 mg 50 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Talabostat mesylate (PT100) is an orally active selective inhibitor of dipeptidyl peptidases IV (DPP4 IC50 0.18 nM) including tumor-associated fibroblast activation protein. This agent may also stimulate the production of colony stimulating factors such as granulocyte colony-stimulating factor (G-CSF) resulting in the stimulation of hematopoiesis. purity: 99%
Catalog No. 50-257-9990