Learn More
Apexbio Technology LLC Talabostat mesylate, 10MM (IN 1ML DMSO) Cas No: 150080-09-4 MFCD: MFCD13194906
SDP
Supplier:  Apexbio Technology LLC B39415.1
Encompass_Preferred
IC50: 1 nMTalabostat mesilate (PT-100; Val-boroPro) is an orally active, specific inhibitor of dipeptidyl peptidases for DPP4, including tumor-associated fibroblast activation protein- (FAP). There are other sizes available. Please inqury us for quote.
Catalog No. NC1658293