Learn More
Medchemexpress LLC HY-15079 5mg Medchemexpress, Talampanel CAS:161832-65-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY150795MG
Encompass_Preferred
Medchemexpress, HY-15079 5mg Talampanel CAS:161832-65-1 Talampanel (LY300164) is an orally and selective α-amino-3-hydroxy-5-methyl-4-isoxazolepropionate (AMPA) receptor antagonis with anti-seizure activity. Talampanel (IVAX) has neuroprotective effects in rodent stroke models. Talampanel attenuates caspase-3 dependent apoptosis in mouse brain. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1679