Learn More
Medchemexpress LLC Talaporfin sodium | 220201-34-3 | 99.7% | 799.69 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY1647750MG
Encompass_Preferred
Talaporfin is a chlorin based photosensitizer. It can be used for the research of various cancers by using photodynamic therapy (PDT).
  • Chlorin based photosensitizer
  • Used for research of various cancers by photodynamic therapy (PDT)
Catalog No. 50-003-94152