Learn More
Cayman Chemical TalaporfInsodIum salt 5mg
SDP
Supplier:  Cayman Chemical 357225 MG
Encompass_Preferred
A photosensitizer; induces apoptosis, DNA DSBs, and the production of ROS in TE-11R 5-fluorouracil-resistant cells at 30 UG/ml in combination with radiation; reduces tumor growth in a TE-11R ESCC mouse xenograft model at 2.5, 5, and 10 mg/kg in combination with PDT
Catalog No. 50-291-1252