Learn More
Cayman Chemical Talaromyceson A 5mg
SDP
Supplier:  Cayman Chemical 329325 MG
Encompass_Preferred
A fungal metabolite; active against S. epidermidis and MRSA (IC50s = 3.70 and 5.48 µM, respectively); inhibits human AChE in a cell-free assay (IC50 = 7.49 µM)
Catalog No. 50-294-8009